Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Triethylsilyl)acetylene, 97%

Catalog No. AAL2021703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2021703 1 g
AAL2021706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2021703 Supplier Thermo Scientific Chemicals Supplier No. L2021703
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 1777-03-3 | C8H16Si | 140.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1777-03-3
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.30
MDL Number MFCD00075373
InChI Key FWSPXZXVNVQHIF-UHFFFAOYSA-N
Synonym triethylsilyl acetylene, ethynyltriethylsilane, triethyl ethynyl silane, triethylsilylacetylene, silane, triethylethynyl, triethylsilyl acetylen, triethylsilyl-acetylene, triethyl-ethynyl-silane
PubChem CID 637947
IUPAC Name triethyl(ethynyl)silane
SMILES CC[Si](CC)(CC)C#C

Specifications

Boiling Point 136°C to 140°C
Quantity 1 g
UN Number UN1993
Beilstein 1743814
Flash Point 17°C (62°F)
Formula Weight 140.31
Percent Purity 97%
Refractive Index 1.4315
Density 0.783
Chemical Name or Material (Triethylsilyl)acetylene
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.