Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethylsilyl Trifluoromethanesulfonate

Catalog No. 503081471 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1471 50 g
50-308-1428 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 50-308-1471 Supplier Sigma Aldrich Supplier No. 27947150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylsilylating agent.

Specifications

CAS 79271-56-0
Density 1.169 g/mL at 25°C
Boiling Point 85°C to 86°C (12 mmHg, lit.)
Molecular Formula C7H15F3O3SSi
Refractive Index n20/D 1.389
Linear Formula CF3SO3Si(C2H5)3
MDL Number MFCD00000407
Quantity 50 g
Synonym TES triflate; Trifluoromethanesulfonic acid triethylsilylester
Molecular Weight (g/mol) 264.34 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.