Learn More
Strem, An Ascensus Company CAS# 1777-03-3. 25g. Triethylsilylacetylene, min. 97%. MFCD00075373
SDP
Catalog No. 501385344
Encompass
CAS# 1777-03-3. 25g. Triethylsilylacetylene, min. 97%. MFCD00075373. Molecular Weight: 140.30. Molecular Formula: (C2H5)3SiC≡CH. Color/form: colorless liq. Strem# 14-7850. http://www.strem.com/catalog/v/14-7850/
Catalog No. 50-138-5344 Supplier Strem, An Ascensus Company Supplier No. 14785025G
CAS# 1777-03-3. 25g. Triethylsilylacetylene, min. 97%. MFCD00075373. Molecular Weight: 140.30. Molecular Formula: (C2H5)3SiC≡CH. Color/form: colorless liq. Strem# 14-7850. http://www.strem.com/catalog/v/14-7850/