Learn More
Medchemexpress LLC Trifarotene | 895542-09-3 | MFCD30481310 | 99.1% | 459.58 g/mol | C29H33NO4 | 5 MG
SDP
Catalog No. 502029451
Encompass_Preferred
Trifarotene is a potent, selective retinoic acid receptor gamma (RARγ) agonist supplied as a solid for research use. It shows strong selectivity for RARγ versus RARα and RARβ, and is provided with supporting documentation for quality and safety.
Catalog No. 50-202-9451 Supplier Medchemexpress LLC Supplier No. HY1002565MG
Trifarotene is a potent, selective retinoic acid receptor gamma (RARγ) agonist supplied as a solid for research use. It shows strong selectivity for RARγ versus RARα and RARβ, and is provided with supporting documentation for quality and safety.