Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trifarotene | 895542-09-3 | 99.1% | 459.58 g/mol | C29H33NO4 | 100 MG
SDP

Catalog No. 5000215595
Encompass_Preferred

Trifarotene is a selective retinoic acid receptor γ (RARγ) agonist supplied as a white to off-white solid for research use. It is intended for in vitro and in vivo studies of RAR/RXR signaling and related pathways and is not for human therapeutic use.

  • High purity (≈99.1% by LCMS)
  • Chemical formula C29H33NO4, molecular weight 459.58 g/mol
  • Soluble in DMSO (≈250 mg/mL) and formulatable for in vivo dosing
  • Stable under recommended powder and solution storage conditions
  • Suitable for receptor pharmacology and preclinical studies

Catalog No. 50-002-15595 Supplier Medchemexpress LLC Supplier No. HY100256100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trifarotene is a selective retinoic acid receptor γ (RARγ) agonist supplied as a white to off-white solid for research use. It is intended for in vitro and in vivo studies of RAR/RXR signaling and related pathways and is not for human therapeutic use.

  • High purity (≈99.1% by LCMS)
  • Chemical formula C29H33NO4, molecular weight 459.58 g/mol
  • Soluble in DMSO (≈250 mg/mL) and formulatable for in vivo dosing
  • Stable under recommended powder and solution storage conditions
  • Suitable for receptor pharmacology and preclinical studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.