Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trifarotene | 895542-09-3 | MFCD30481310 | 99.1% | 459.58 | C29H33NO4 | 50 MG
SDP

Catalog No. 5000215408
Encompass_Preferred

Trifarotene is a synthetic retinoid supplied for research use as a potent and selective retinoic acid receptor γ (RARγ) agonist. Provided as a solid for in vitro and analytical applications, it is intended for laboratory research and is not for human or clinical use.

  • High purity (99.12%).
  • Molecular formula C29H33NO4.
  • Molecular weight 459.58 g/mol.
  • Selective RARγ agonist (EC50 = 7.7 nM) with ~65-fold and ~16-fold selectivity versus RARα and RARβ.
  • Supplied as 50 mg quantity suitable for analytical and experimental workflows.
  • Intended for research and analytical applications only; not for human use.

Catalog No. 50-002-15408 Supplier Medchemexpress LLC Supplier No. HY10025650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trifarotene is a synthetic retinoid supplied for research use as a potent and selective retinoic acid receptor γ (RARγ) agonist. Provided as a solid for in vitro and analytical applications, it is intended for laboratory research and is not for human or clinical use.

  • High purity (99.12%).
  • Molecular formula C29H33NO4.
  • Molecular weight 459.58 g/mol.
  • Selective RARγ agonist (EC50 = 7.7 nM) with ~65-fold and ~16-fold selectivity versus RARα and RARβ.
  • Supplied as 50 mg quantity suitable for analytical and experimental workflows.
  • Intended for research and analytical applications only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.