Learn More
Med Vet International Trifexis Blue, 40.1-60Lb, 6 Dose
Catalog No. 502828338
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
PRODUCT DETAILS Trifexis is a monthly chewable tablet that provides protection against three types of parasites for your dog. Trifexis: 1) Kills fleas and prevents infestations 2) Prevents heartworm disease 3) Treats and controls adult hookworm, roundworm and whipworm infections. Trifexis is a beef flavored tablet that starts killing fleas in 30 minutes. With Trifexis, there is no need to isolate the dog from other pets or children. Pet owners can play with their dog immediately after treatment. Trifexis is approved for use in dogs and puppies 8 weeks of age or older and 5 pounds of body weight or greater
Catalog No. 50-282-8338 Supplier Med Vet International Supplier No. RXTRIFEXISBL
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
PRODUCT DETAILS Trifexis is a monthly chewable tablet that provides protection against three types of parasites for your dog. Trifexis: 1) Kills fleas and prevents infestations 2) Prevents heartworm disease 3) Treats and controls adult hookworm, roundworm and whipworm infections. Trifexis is a beef flavored tablet that starts killing fleas in 30 minutes. With Trifexis, there is no need to isolate the dog from other pets or children. Pet owners can play with their dog immediately after treatment. Trifexis is approved for use in dogs and puppies 8 weeks of age or older and 5 pounds of body weight or greater