Learn More
Mayflower Bioscience TRIFic CD9 Exosome Assay, 96 well
SDP
Catalog No. 501979304
Encompass
The TRIFic™ exosome detection assay delivers clear and consistent data quickly by providing quantitative data from purified or unpurified samples, including direct measurement of exosomes in plasma in a convenient 96-well format. It is similar to an ELISA assay, but the target is detected directly with a Europium label without any enzymatic reaction. TRIFic™ exosome detection assays are being made available for widely used exosome markers, allowing for identification of CD9+/CD81+/CD63+ - EVs.
Catalog No. 50-197-9304 Supplier Mayflower Bioscience Supplier No. EX501
The TRIFic™ exosome detection assay delivers clear and consistent data quickly by providing quantitative data from purified or unpurified samples, including direct measurement of exosomes in plasma in a convenient 96-well format. It is similar to an ELISA assay, but the target is detected directly with a Europium label without any enzymatic reaction. TRIFic™ exosome detection assays are being made available for widely used exosome markers, allowing for identification of CD9+/CD81+/CD63+ - EVs.