Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trifloxystrobin | 141517-21-7 | MFCD09953763 | 99.2% | 408.37 g/mol | C20H19F3N2O4 | 10MG
SDP

Catalog No. 5000239024
Encompass_Preferred

Trifloxystrobin analytical standard supplied as a solid reference material for laboratory analytical use. It is intended for method development, calibration, and verification in HPLC, GC, and MS workflows, and is provided in small pack sizes suitable for routine research applications.

  • High purity reference material for accurate quantitation.
  • Molecular weight 408.37 g/mol and formula C20H19F3N2O4.
  • Suitable for HPLC, GC, and MS method development.
  • Available in small pack sizes including 10 mg for low-consumption assays.
  • Supplied as a solid analytical standard for research use.

Catalog No. 50-002-39024 Supplier Medchemexpress LLC Supplier No. HY123230R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trifloxystrobin analytical standard supplied as a solid reference material for laboratory analytical use. It is intended for method development, calibration, and verification in HPLC, GC, and MS workflows, and is provided in small pack sizes suitable for routine research applications.

  • High purity reference material for accurate quantitation.
  • Molecular weight 408.37 g/mol and formula C20H19F3N2O4.
  • Suitable for HPLC, GC, and MS method development.
  • Available in small pack sizes including 10 mg for low-consumption assays.
  • Supplied as a solid analytical standard for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.