Learn More
Medchemexpress LLC Trifloxystrobin | 141517-21-7 | MFCD09953763 | 99.2% | 408.37 g/mol | C20H19F3N2O4 | 25MG
SDP
Catalog No. 5000238602
Encompass_Preferred
Trifloxystrobin (standard) is an analytical reference standard of the fungicide trifloxystrobin supplied for research and analytical applications. Provided as a 25 mg vial with high purity, it is intended for calibration, method validation, and residue analysis. Molecular formula: C20H19F3N2O4; molecular weight: 408.37 g/mol.
  • High purity suitable for analytical use.
  • Intended for calibration and method validation.
  • Supplied in a small quantity appropriate for trace-level analysis.
  • Certificate of analysis and safety data sheet available.
  • Appropriate for environmental and residue testing workflows.
Catalog No. 50-002-38602 Supplier Medchemexpress LLC Supplier No. HY123230R25MG
Add to Cart
Add to Cart
Trifloxystrobin (standard) is an analytical reference standard of the fungicide trifloxystrobin supplied for research and analytical applications. Provided as a 25 mg vial with high purity, it is intended for calibration, method validation, and residue analysis. Molecular formula: C20H19F3N2O4; molecular weight: 408.37 g/mol.
  • High purity suitable for analytical use.
  • Intended for calibration and method validation.
  • Supplied in a small quantity appropriate for trace-level analysis.
  • Certificate of analysis and safety data sheet available.
  • Appropriate for environmental and residue testing workflows.