Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trifloxystrobin | 141517-21-7 | MFCD09953763 | 99.2% | 408.37 g/mol | C20H19F3N2O4 | 5MG
SDP

Catalog No. 5000239150
Encompass_Preferred

Trifloxystrobin analytical standard is a strobilurin-class fungicide supplied as a white to off-white solid for research and analytical applications. The material is provided as a high-purity reference standard for method development, calibration, and compound characterization; see manufacturer documentation for storage and handling details.

  • High purity 99.2%.
  • Molecular weight 408.37 g/mol.
  • Chemical formula C20H19F3N2O4.
  • White to off-white solid appearance.
  • Supplied in small reference sizes suitable for analytical use.
  • Intended for method development and quantification of the compound.

Catalog No. 50-002-39150 Supplier Medchemexpress LLC Supplier No. HY123230R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trifloxystrobin analytical standard is a strobilurin-class fungicide supplied as a white to off-white solid for research and analytical applications. The material is provided as a high-purity reference standard for method development, calibration, and compound characterization; see manufacturer documentation for storage and handling details.

  • High purity 99.2%.
  • Molecular weight 408.37 g/mol.
  • Chemical formula C20H19F3N2O4.
  • White to off-white solid appearance.
  • Supplied in small reference sizes suitable for analytical use.
  • Intended for method development and quantification of the compound.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.