Learn More
Cayman Chemical TrIfloxystrobIn 50mg
SDP
Catalog No. 502934333
Encompass_Preferred
A fungicide; inhibits the growth of R. solani isolates from sugar beet crops (EC50s = 0.14-823.54 μg/ml); inhibits mycelial growth of C. gloeosporioides when used in combination with tebuconazole from 100-1,000 ppm; toxic to various aquatic species, including R. arenarum, P. santafecinus, E. bicolor, and L. latrans tadpoles (LC50s = 0.22, 0.14, 0.1, and 0.26 mg/L, respectively); reduces mobility of tadpoles and decreases predation of trifloxystrobin-exposed, but not non-exposed, tadpoles by eels
Catalog No. 50-293-4333 Supplier Cayman Chemical Supplier No. 2582550 MG
A fungicide; inhibits the growth of R. solani isolates from sugar beet crops (EC50s = 0.14-823.54 μg/ml); inhibits mycelial growth of C. gloeosporioides when used in combination with tebuconazole from 100-1,000 ppm; toxic to various aquatic species, including R. arenarum, P. santafecinus, E. bicolor, and L. latrans tadpoles (LC50s = 0.22, 0.14, 0.1, and 0.26 mg/L, respectively); reduces mobility of tadpoles and decreases predation of trifloxystrobin-exposed, but not non-exposed, tadpoles by eels