Learn More
FUJIFILM Biosciences Trifloxysulfuron Standard
Catalog No. 5000450345
Encompass_Preferred
A high-purity Trifloxysulfuron reference standard 98 0% HPLC for use in pesticide residue analysis Trifloxysulfuron is a sulfonylurea herbicide Suitable for laboratory research and regulated testing environments Supplied as a white to nearly white powder
Catalog No. 50-004-50345 Supplier FUJIFILM Biosciences Supplier No. 20117861
A high-purity Trifloxysulfuron reference standard 98 0% HPLC for use in pesticide residue analysis Trifloxysulfuron is a sulfonylurea herbicide Suitable for laboratory research and regulated testing environments Supplied as a white to nearly white powder