Learn More
Chem Service Inc Triflumuron, 64628-44-0, MFCD00072496, 100 mg
SDP
Catalog No. NC1528427
Encompass_Preferred
Molecular Formula: C15H10ClF3N2O3, Molecular Weight: 358.72, Classification: Insecticide. Triflumuron is a non-sysmetic insect growth regulator or insecticide, which can inhibit chitin synthesis. Its can act against moulting the larvae and also cause infertility of the eggs.
Catalog No. NC1528427 Supplier Chem Service Inc Supplier No. N13687100MG
Molecular Formula: C15H10ClF3N2O3, Molecular Weight: 358.72, Classification: Insecticide. Triflumuron is a non-sysmetic insect growth regulator or insecticide, which can inhibit chitin synthesis. Its can act against moulting the larvae and also cause infertility of the eggs.