Learn More
Medchemexpress LLC Triflumuron | 64628-44-0 | 99.0% | 10 G
SDP
Catalog No. 5000411627
Encompass_Preferred
Triflumuron is a potent chitin synthesis inhibitor that can be utilized as an insecticide and represents a potential tool for controlling disease vectors in public health. It demonstrates efficacy in emergence inhibition (EI) against *Culex quinquefasciatus* and *Aedes albopictus*.
  • Potent chitin synthesis inhibitor
  • Effective in emergence inhibition (EI) against *Culex quinquefasciatus* and *Aedes albopictus*
  • Can be used as an insecticide
  • Potential tool for disease vector control in public health
  • For research use only
Catalog No. 50-004-11627 Supplier Medchemexpress LLC Supplier No. HYB204910G
Add to Cart
Add to Cart
Triflumuron is a potent chitin synthesis inhibitor that can be utilized as an insecticide and represents a potential tool for controlling disease vectors in public health. It demonstrates efficacy in emergence inhibition (EI) against *Culex quinquefasciatus* and *Aedes albopictus*.
  • Potent chitin synthesis inhibitor
  • Effective in emergence inhibition (EI) against *Culex quinquefasciatus* and *Aedes albopictus*
  • Can be used as an insecticide
  • Potential tool for disease vector control in public health
  • For research use only