Learn More
Medchemexpress LLC Triflumuron | 64628-44-0 | MFCD00072496 | 99.0% | 25 G
SDP
Catalog No. 5000414266
Encompass_Preferred
Triflumuron is a potent chitin synthesis inhibitor effective against disease vectors like *Culex quinquefasciatus* and *Aedes albopictus*, demonstrating its utility as an insecticide in public health.
  • Potent chitin synthesis inhibitor
  • Effective in emergence inhibition against *Culex quinquefasciatus*
  • Effective in emergence inhibition against *Aedes albopictus*
  • Can be used as an insecticide
  • Potential tool for controlling disease vectors
Catalog No. 50-004-14266 Supplier Medchemexpress LLC Supplier No. 25GHYB2049
Triflumuron is a potent chitin synthesis inhibitor effective against disease vectors like *Culex quinquefasciatus* and *Aedes albopictus*, demonstrating its utility as an insecticide in public health.
  • Potent chitin synthesis inhibitor
  • Effective in emergence inhibition against *Culex quinquefasciatus*
  • Effective in emergence inhibition against *Aedes albopictus*
  • Can be used as an insecticide
  • Potential tool for controlling disease vectors