Learn More
Apexbio Technology LLC Trifluoperazine 2HCl 440-17-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062218
Encompass_Preferred
Trifluoperazine 2HCl is a dopamine D2 receptor antagonist extensively utilized in biomedical research for investigating dopaminergic signaling pathways It functions by selectively inhibiting the D2 subtype dopamine receptor exhibiting an IC50 value of approximately 1 1 nM Due to its receptor-blocking mechanism trifluoperazine is frequently employed to study dopamine-mediated neurotransmission receptor pharmacodynamics and neurological processes underlying disorders involving altered dopaminergic activity such as schizophrenia Additionally trifluoperazine serves as a tool compound for elucidating cellular responses linked to dopaminergic receptor modulation in in vitro receptor-binding assays and neuronal cell cultures
Catalog No. 50-000-62218 Supplier Apexbio Technology LLC Supplier No. B13975.1
Trifluoperazine 2HCl is a dopamine D2 receptor antagonist extensively utilized in biomedical research for investigating dopaminergic signaling pathways It functions by selectively inhibiting the D2 subtype dopamine receptor exhibiting an IC50 value of approximately 1 1 nM Due to its receptor-blocking mechanism trifluoperazine is frequently employed to study dopamine-mediated neurotransmission receptor pharmacodynamics and neurological processes underlying disorders involving altered dopaminergic activity such as schizophrenia Additionally trifluoperazine serves as a tool compound for elucidating cellular responses linked to dopaminergic receptor modulation in in vitro receptor-binding assays and neuronal cell cultures