Learn More
Selleck Chemical LLC Trifluoperazine 2HCl S3201-1g
SDP
Catalog No. 503109985
Encompass_Preferred
Trifluoperazine (NSC-17474 RP-7623 SKF-5019) 2HCl is a dopamine D2 receptor inhibitor with IC50 of 1 1 nM Trifluoperazine also inhibits calmodulin (CaM)
Catalog No. 50-310-9985 Supplier Selleck Chemical LLC Supplier No. S32011G
Trifluoperazine (NSC-17474 RP-7623 SKF-5019) 2HCl is a dopamine D2 receptor inhibitor with IC50 of 1 1 nM Trifluoperazine also inhibits calmodulin (CaM)