Learn More
Selleck Chemical LLC Trifluoperazine 2HCl-50mg
SDP
Supplier:  Selleck Chemical LLC S320150MG
Encompass_Preferred
Trifluoperazine (NSC-17474, RP-7623, SKF-5019) 2HCl is a dopamine D2 receptor inhibitor with IC50 of 1.1 nM. Trifluoperazine also inhibits calmodulin (CaM).
Catalog No. 50-238-9971