Learn More
Lkt Laboratories Trifluoperazine Dihydrochloride, ≥98%, 5GR
SDP
Supplier:  Lkt Laboratories T70335G
Encompass_Preferred
Phenothiazine; D1/2 and α1-adrenergic antagonist, calmodulin and CDPK4 inhibitor, Nav1.4 and Nav1.7 Na+ channel blocker. CAS: 440-17-5. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. NC0017620