Learn More
Enzo Life Sciences Trifluoperazine . dihydrochloride (5g). CAS: 440-17-5
SDP
Supplier:  Enzo Life Sciences ALX550310G005
Encompass_Preferred
Facilitates agonist induced elevation of cytosolic calcium at low concentrations (10µM). Antagonizes calmodulin at higher concentrations. Antipsychotic and sedative dopamine antagonist. Potent and irreversible inhibitor of cAMP-gated cationic channels. Inhibits DNA repair. Cell permeable. Potent neurotoxin. Purity: ≥98% (UHPLC). Solubility: Soluble in water (50mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9532