Learn More
Abcam Trifluoperazine dihydrochloride, calmodulin antagonist, 1G
Catalog No. 502707316
Encompass_Preferred
MW 480.42 g/mol, Purity >99%. Cell-permeable calmodulin antagonist. Dopamine receptor antagonist. Stimulates ER Ca²+ ATPase activity at low concentrations and inhibits at higher concentrations. Antipsychotic.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7316 Supplier Abcam Supplier No. AB1207391G
MW 480.42 g/mol, Purity >99%. Cell-permeable calmodulin antagonist. Dopamine receptor antagonist. Stimulates ER Ca²+ ATPase activity at low concentrations and inhibits at higher concentrations. Antipsychotic.
The product is subject to the following: Abcam Restricted Use Statement