Learn More
Medchemexpress LLC Trifluoperazine dihydrochloride | 440-17-5 | 99.9% | 100 MG
SDP
Catalog No. 5000405849
Encompass_Preferred
Trifluoperazine dihydrochloride is a potent antipsychotic agent that functions by blocking central dopamine receptors. This compound also acts as a strong α1-adrenergic receptor antagonist and a NUPR1 inhibitor, demonstrating anticancer activity. Additionally, it is a calmodulin inhibitor and can inhibit P-glycoprotein. Trifluoperazine dihydrochloride is suitable for research related to schizophrenia and has been shown to be a reversible inhibitor of influenza virus morphogenesis.
  • Potent antipsychotic agent
  • Blocks central dopamine receptors
  • Strong α1-adrenergic receptor antagonist
  • NUPR1 inhibitor with anticancer activity
  • Calmodulin inhibitor
  • Inhibits P-glycoprotein
  • Suitable for schizophrenia research
  • Reversible inhibitor of influenza virus morphogenesis
  • IC50 of 16 μM for cytotoxicity in HL-60 cells by MTT assay
Catalog No. 50-004-05849 Supplier Medchemexpress LLC Supplier No. HYB0532A100MG
Trifluoperazine dihydrochloride is a potent antipsychotic agent that functions by blocking central dopamine receptors. This compound also acts as a strong α1-adrenergic receptor antagonist and a NUPR1 inhibitor, demonstrating anticancer activity. Additionally, it is a calmodulin inhibitor and can inhibit P-glycoprotein. Trifluoperazine dihydrochloride is suitable for research related to schizophrenia and has been shown to be a reversible inhibitor of influenza virus morphogenesis.
  • Potent antipsychotic agent
  • Blocks central dopamine receptors
  • Strong α1-adrenergic receptor antagonist
  • NUPR1 inhibitor with anticancer activity
  • Calmodulin inhibitor
  • Inhibits P-glycoprotein
  • Suitable for schizophrenia research
  • Reversible inhibitor of influenza virus morphogenesis
  • IC50 of 16 μM for cytotoxicity in HL-60 cells by MTT assay