Learn More
Medchemexpress LLC Trifluoperazine dihydrochloride | 440-17-5 | 99.9% | 50 MG
SDP
Catalog No. 5000407614
Encompass_Preferred
Trifluoperazine dihydrochloride is an antipsychotic agent that functions by blocking central dopamine receptors. It also acts as a potent α1-adrenergic receptor antagonist and a NUPR1 inhibitor, demonstrating anticancer activity. Furthermore, it is a calmodulin inhibitor and inhibits P-glycoprotein. This compound is utilized in research for schizophrenia and acts as a reversible inhibitor of influenza virus morphogenesis by interfering with cellular CaM and/or CaM-dependent functions.
  • Blocks central dopamine receptors
  • Potent α1-adrenergic receptor antagonist
  • Potent NUPR1 inhibitor with anticancer activity
  • Calmodulin inhibitor
  • Inhibits P-glycoprotein
  • Utilized in research for schizophrenia
  • Acts as a reversible inhibitor of influenza virus morphogenesis
Catalog No. 50-004-07614 Supplier Medchemexpress LLC Supplier No. HYB0532A50MG
Trifluoperazine dihydrochloride is an antipsychotic agent that functions by blocking central dopamine receptors. It also acts as a potent α1-adrenergic receptor antagonist and a NUPR1 inhibitor, demonstrating anticancer activity. Furthermore, it is a calmodulin inhibitor and inhibits P-glycoprotein. This compound is utilized in research for schizophrenia and acts as a reversible inhibitor of influenza virus morphogenesis by interfering with cellular CaM and/or CaM-dependent functions.
  • Blocks central dopamine receptors
  • Potent α1-adrenergic receptor antagonist
  • Potent NUPR1 inhibitor with anticancer activity
  • Calmodulin inhibitor
  • Inhibits P-glycoprotein
  • Utilized in research for schizophrenia
  • Acts as a reversible inhibitor of influenza virus morphogenesis