Learn More
Medchemexpress LLC Trifluoperazine dihydrochloride | 440-17-5 | 99.98% | 10 MM 1 ML
SDP
Catalog No. 5000406874
Encompass_Preferred
Trifluoperazine dihydrochloride is an antipsychotic agent that functions by blocking central dopamine receptors. It also acts as a potent alpha1-adrenergic receptor antagonist and a NUPR1 inhibitor, exhibiting anticancer activity. Furthermore, it is identified as a calmodulin inhibitor and an inhibitor of P-glycoprotein. This compound is suitable for research related to schizophrenia and has been observed to act as a reversible inhibitor of influenza virus morphogenesis.
  • Antipsychotic agent
  • Blocks central dopamine receptors
  • Potent alpha1-adrenergic receptor antagonist
  • Potent NUPR1 inhibitor
  • Exhibits anticancer activity
  • Calmodulin inhibitor
  • Inhibitor of P-glycoprotein
  • Suitable for research related to schizophrenia
  • Reversible inhibitor of influenza virus morphogenesis
Catalog No. 50-004-06874 Supplier Medchemexpress LLC Supplier No. HYB0532A10MM/1ML
Trifluoperazine dihydrochloride is an antipsychotic agent that functions by blocking central dopamine receptors. It also acts as a potent alpha1-adrenergic receptor antagonist and a NUPR1 inhibitor, exhibiting anticancer activity. Furthermore, it is identified as a calmodulin inhibitor and an inhibitor of P-glycoprotein. This compound is suitable for research related to schizophrenia and has been observed to act as a reversible inhibitor of influenza virus morphogenesis.
  • Antipsychotic agent
  • Blocks central dopamine receptors
  • Potent alpha1-adrenergic receptor antagonist
  • Potent NUPR1 inhibitor
  • Exhibits anticancer activity
  • Calmodulin inhibitor
  • Inhibitor of P-glycoprotein
  • Suitable for research related to schizophrenia
  • Reversible inhibitor of influenza virus morphogenesis