Learn More
Abcam 4-Trifluoromethylumbelliferyl oleate, 10MG
Catalog No. 50996112
Encompass_Preferred
MW 494.6 Da. Fluorescent lipase substrate. Releases highly fluorescent 4-trifluoromethylumbelliferone upon hydrolysis. 4-trifluoromethylumbelliferone is insensitive to pH making it suitable for use in food analysis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-112 Supplier Abcam Supplier No. AB27507210MG
MW 494.6 Da. Fluorescent lipase substrate. Releases highly fluorescent 4-trifluoromethylumbelliferone upon hydrolysis. 4-trifluoromethylumbelliferone is insensitive to pH making it suitable for use in food analysis.
The product is subject to the following: Abcam Restricted Use Statement