Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid 2,2,2-trifluoroethylester 99+%, Thermo Scientific™

Catalog No. AC243420250
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC243420250 25 mL
AC243421000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC243420250 Supplier Thermo Scientific Chemicals Supplier No. 243420250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-38-5
Molecular Formula C4H2F6O2
Molecular Weight (g/mol) 196.05
MDL Number MFCD00000418
InChI Key ZKUJOCJJXCPCFS-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl trifluoroacetate, acetic acid, trifluoro-, 2,2,2-trifluoroethyl ester, trifluoroacetic acid 2,2,2-trifluoroethyl ester, 2,2,2-trifluoroethyltrifluoroacetate, pubchem12623, acmc-1ake7, cf3co2ch2cf3, cf3c o och2cf3, ksc496e0r, trifluoroethyl trifluoroacetate
PubChem CID 67888
IUPAC Name 2,2,2-trifluoroethyl 2,2,2-trifluoroacetate
SMILES FC(F)(F)COC(=O)C(F)(F)F

Specifications

Color Colorless
Density 1.46
Infrared Spectrum Authentic
Refractive Index 1.2745 to 1.2765 (20°C, 589nm)
Linear Formula CF3CO2CH2CF3
Quantity 25 mL
Beilstein 02, 427
Fieser 16357
Formula Weight 196.05
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material Trifluoroacetic acid 2, 2, 2-trifluoroethylester, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 206-985-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.