Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020564 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0564 1 mL
11-101-9696 5 mL
11-101-8847 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-0564 Supplier MilliporeSigma Supelco Supplier No. 745641MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trifluoroacetic acid is a phytotoxic compound belonging to the class of perfluorinated acids (PFAs). It is an environmental contaminant, which contributes to the organo-fluorine burden in surface and ground waters.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 76-05-1
Vapor Pressure 97.5 mmHg (20°C)
Vapor Density 3.9 (vs. air)
Density 1.489 g/mL (at 20°C)
Physical Form Neat
Linear Formula CF3 COOH
MDL Number MFCD00004169
Melting Point -15.4°C
Boiling Point 72.4°C
Synonym TFA
Solubility Soluble in water
Type Analytical Standard
Quantity 1 mL
Formula Weight 114.02
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.