Learn More
Strem, An Ascensus Company CAS# 76-05-1. 500g. Trifluoroacetic acid, 99%. MFCD00004169
SDP
Catalog No. 5090109424
Encompass
CAS# 76-05-1. 500g. Trifluoroacetic acid, 99%. MFCD00004169. Molecular Weight: 114.02. Molecular Formula: CF3COOH. Color/form: colorless liq. Strem# 09-7160. http://www.strem.com/catalog/v/09-7160/
Catalog No. 50-901-09424 Supplier Strem, An Ascensus Company Supplier No. 097160500G
CAS# 76-05-1. 500g. Trifluoroacetic acid, 99%. MFCD00004169. Molecular Weight: 114.02. Molecular Formula: CF3COOH. Color/form: colorless liq. Strem# 09-7160. http://www.strem.com/catalog/v/09-7160/