Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, BioSyn™, for biosynthesis, for HPLC, >99.5%, Solstice

Catalog No. 60048603 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
12 x 50 mL
6 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-603 6 x 500 mL
60-048-602 12 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-048-603 Supplier Solstice Supplier No. BB360P500

missing translation for 'validMainframeMsg'

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Boiling Point 72°C
CAS Max % 100.00
Grade HPLC
Density 1.48 g/cm3
Quantity 6 x 500 mL
Packaging Glass Bottle
Physical Form Liquid
Hazard Category Danger
Hazard Statement H332+H314+H290
Precautionary Statement P280+P234+P261+P264+P271
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.