Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, CHROMASOLV™, for HPLC, ≥99.0%, Solstice

Catalog No. 60045808 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-808 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-808 Supplier Solstice Supplier No. 3020311L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ for HPLC,≥99.0%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Absorbance max. 0.10 at 270 nm, max. 0.9 at 260 nm, max. 0.025 at 320 nm, max. 0.03 at 300 nm, max. 0.04 at 290 nm, max. 0.05 280 nm
Melting Point -15.4°C (literature)
Boiling Point 72°C
CAS Max % 100.0000%
Density 1.489 g/mL at 20 °C(lit.)
Quantity 1 L
Flash Point 100°C
Packaging Glass Bottle
Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
Formula Weight 114.02
Percent Purity ≥99%
Grade HPLC
Chemical Name or Material Trifluoroacetic acid, CHROMASOLV™, for HPLC, ≥99.0%
Show More Show Less
ShelfLife 360 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: I; Proper Shipping Name: Trifluoroacetic acid
EINECSNumber 200-929-3
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.