Learn More
Sigma Aldrich Fine Chemicals Biosciences Trifluoroacetic acid-d 99.25G
Catalog No. 502899717
Encompass
Trifluoroacetic acid-d is a synthetic reagent having isotopic purity of 99.5atom%D. It undergoes SEAE reaction with (R)-(E)-1 3-diphenyl-3-trimethylsilylpropene via antistereochemistry to afford (R)-(E)-1 3-diphenylpropene-3-d. Vapor phase dissociation of trifluoroacetic acid-d has been studied.
Catalog No. 50-289-9717 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 15200525G
Trifluoroacetic acid-d is a synthetic reagent having isotopic purity of 99.5atom%D. It undergoes SEAE reaction with (R)-(E)-1 3-diphenyl-3-trimethylsilylpropene via antistereochemistry to afford (R)-(E)-1 3-diphenylpropene-3-d. Vapor phase dissociation of trifluoroacetic acid-d has been studied.