Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid-d, 99.5%(Isotopic)

Catalog No. AA4235709
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4235709 10 g
AA4235714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4235709 Supplier Thermo Scientific Chemicals Supplier No. 04235709
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 599-00-8 | C2HF3O2 | 115.029 g/mol

Trifluoroacetic acid-d is employed as solvent to investigate the conformations of cyclic peptides by proton NMR spectra. It is also used to compose NMR solvent mixture to investigate the NMR spectra of high molecular weight polyesters such as polyglycolide (PG) and poly(ethylene terephthalate)and as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-00-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 115.029
MDL Number MFCD00044563
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
Synonym trifluoroacetic acid-d, deuterotrifluoroacetic acid, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, acetic acid-d, 2,2,2-trifluoro, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d, tfa-d, cf3cood, trifluoroacetic 2h acid
PubChem CID 71502
IUPAC Name deuterio 2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)O

Specifications

Physical Form Liquid
Quantity 10 g
Packaging Prescored Ampule
Density 1.50 g/mL
Odor Acrid
Sensitivity Hygroscopic
Melting Point -15°C
Boiling Point 72°C
UN Number UN2699
Solubility Information Soluble in water.
Formula Weight 115.03
Percent Purity 99.5%
Chemical Name or Material Trifluoroacetic acid-d, Isotopic
Show More Show Less
Hazard Category H290-H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: TRIFLUOROACETIC ACID
EINECSNumber 209-961-2
TSCA Yes
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.