Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid-d, for NMR, 99 atom % D

Catalog No. AC166400050
Change view
Click to view available options
Quantity:
5 mL
10 mL
25 mL
100 mL
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166400050 5 mL Glass Bottle
AC166400100 10 mL Glass Bottle
AC166400250 25 mL Glass Bottle
AC166401000 100 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC166400050 Supplier Thermo Scientific Chemicals Supplier No. 166400050
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 599-00-8 | C2HF3O2 | 115.029 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-00-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 115.029
MDL Number MFCD00044563
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
PubChem CID 71502
IUPAC Name deuterio 2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)O

Specifications

Physical Form Liquid
Color Colorless
Packaging Glass Bottle
Quantity 5 mL
Density 1.5000g/mL
Water 0.1% max. (Coulometric)
Deuterium (D) 98.5% min.
Melting Point -15.0°C
Boiling Point 75.0°C
Linear Formula CF3CO2D
Synonym trifluoroacetic acid-d, deuterotrifluoroacetic acid, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, acetic acid-d, 2,2,2-trifluoro, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d, tfa-d, cf3cood, trifluoroacetic 2h acid
Formula Weight 115.03
Grade NMR
Chemical Name or Material Trifluoroacetic acid-d, 99 atom%D
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.