Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid-D1, deuteration degree min. 99.5% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Catalog No. M1133630010 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1133630010 10 mL Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1133630010 Supplier MilliporeSigma Supplier No. 1.13363.0010
Add to Cart
Add to Cart

For NMR Spectroscopy

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-00-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 115.029
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
Synonym trifluoroacetic acid-d, deuterotrifluoroacetic acid, acetic acid-d, trifluoro, acetic acid-d,2,2,2-trifluoro, acetic acid-d, 2,2,2-trifluoro, trifluoroacetic acid-d tfa-d, trifluoroacetic acid-d, 99.5 atom % d, tfa-d, cf3cood, trifluoroacetic 2h acid
PubChem CID 71502
IUPAC Name deuterio 2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)O

Specifications

Physical Form Liquid
Quantity 10 mL
Packaging Glass Ampule
Odor Stinging
Assay Percent Range 99.5% min.
Formula Weight 115.03
Chemical Name or Material Trifluoroacetic acid-D1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.