Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, For protein sequencing, Solstice

Catalog No. 6001756 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-017-56 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-017-56 Supplier Solstice Supplier No. 299537500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purified by redistillation, for protein sequencing,≥99%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Density 1.489g/mL at 20°C
Quantity 500 g
Flash Point >100°C (>212°F)
Linear Formula CF3COOH
Vapor Density 3.9 (vs air)
UN Number 2699
Vapor Pressure 97.5mmHg (20°C)
Beilstein 742035
Formula Weight 114.02g/mol
Percent Purity ≥99%
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P310
Precautionary Statement H314-H332-H412
EINECSNumber 200-929-3
RTECSNumber AJ9625000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.