Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, Solstice

Catalog No. 60048615 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Packaging:
Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-615 10 x 1 mL Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-615 Supplier Solstice Supplier No. 4096710X1ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
PubChem CID 6422
ChEBI CHEBI:45892

Specifications

Percent Purity 99.0% (min.)
Grade LC-MS Eluents
Quantity 10 x 1 mL
Flash Point 100 °C
Packaging Ampoule
Physical Form Liquid
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.