Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, HPLC Grade, J.T. Baker™

Catalog No. 02004498 Shop All Avantor Products
Change view
Click to view available options
Quantity:
10 x 1 mL
900 mL
Packaging:
Glass Ampoule
Narrow Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-498 900 mL Narrow Mouth Amber Glass Bottle
02-004-494 10 x 1 mL Glass Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-004-498 Supplier Avantor Supplier No. 947002

missing translation for 'validMainframeMsg'

High purity solvents for use in analytical and preparative separations

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Formula Weight 114.03
Grade HPLC
Quantity 900 mL
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.53kg
Chemical Name or Material Trifluoroacetic Acid, Gradient Tested for HPLC Peptide and Protein Analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.