Learn More
Covachem LCMS Grade TFA (Trifluoroacetic acid) 25ml
Catalog No. NC3717738
Encompass
CovaChem's LCMS Grade Trifluoroacetic acid (TFA) is well suited as a chromatography additive for difficult to separate analytes in HPLC and LCMS applications. CovaChem's LCMS TFA is an ideal additive for separations of peptides in peptide synthesis applications. Trifluoroacetic acid is widely used to separate both protected amino acid chains and deprotected peptides. Due to the amino and carboxylate groups having vastly different pka values, synthetic peptides can be particularly challenging to separate, The HPLC separation of peptides is much more easily achieved with 0.1% Trifluoroacetic acid being present in the mobile phase.
Catalog No. NC3717738 Supplier Covachem Supplier No. 1120425
CovaChem's LCMS Grade Trifluoroacetic acid (TFA) is well suited as a chromatography additive for difficult to separate analytes in HPLC and LCMS applications. CovaChem's LCMS TFA is an ideal additive for separations of peptides in peptide synthesis applications. Trifluoroacetic acid is widely used to separate both protected amino acid chains and deprotected peptides. Due to the amino and carboxylate groups having vastly different pka values, synthetic peptides can be particularly challenging to separate, The HPLC separation of peptides is much more easily achieved with 0.1% Trifluoroacetic acid being present in the mobile phase.