Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProteoSpec™ Trifluoroacetic Acid, LCMS Grade, Ricca Chemical
SDP

Catalog No. RMB11204101 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Packaging:
Glass Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RMB11204101 10 x 1 mL Glass Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. RMB11204101 Supplier Ricca Chemical Company Supplier No. RMB1120410X1R

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LCMS Mobile Phase

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

pH <2
Melting Point -15°C
Boiling Point 76°C
CAS Min % 100
Color Colorless
CAS Max % 100
Solubility Information Miscible in water
Formula Weight 114.02
Grade LC-MS
Density 1.535
Quantity 10 x 1 mL
Packaging Glass Ampule
Physical Form Liquid
Chemical Name or Material ProteoSpec™ Trifluoroacetic Acid, LCMS Grade
Show More Show Less
ShelfLife 12 months
Recommended Storage Ambient Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.