Learn More
Description
- Extensive QC testing ensures the highest specifications
- Optimized to improve ionization and resolution
- Extremely low levels of inorganic and organic impurities
- Manufactured specifically for accurate and fast LC-MS
Specifications
Specifications
| Melting Point | -15.4°C (literature) |
| Boiling Point | 72.4°C (literature) |
| Density | 1.489 g/mL (at 20°C (literature)) |
| Quantity | 10 x 1 mL |
| Refractive Index | n20/D 1.3 (literature) |
| Linear Formula | CF3COOH |
| Vapor Density | 3.9 (vs air) |
| Vapor Pressure | 97.5 mmHg (20°C) |
| UN Number | UN 2699 8/PGI |
| Formula Weight | 114.02 |
| Show More |
Safety and Handling
| Hazard Category | H314 - H332 - H412 |
| RTECSNumber | AJ9625000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.