Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, Reagent Grade, 99%, Solstice

Catalog No. 6001759 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
10 Ampules
100 mL
25 mL
5 x 10 Ampules
500 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-017-59 1 L
60-017-60 25 mL
60-017-58 100 mL
60-017-61 500 mL
60-048-201 10 Ampules
60-048-203 5 x 10 Ampules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-017-59 Supplier Solstice Supplier No. T65081L
Add to Cart
Add to Cart

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
Formula Weight 114.02g/mol
Percent Purity 99%
Grade ReagentPlus
Density 1.489g/mL at 20°C
Quantity 1 L
Flash Point >100°C (>212°F)
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
Hazard Statement P261-P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
RTECSNumber AJ9625000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.