Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic Acid, Reagents

Catalog No. 01336907
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-907 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-907 Supplier Reagents Holdings Llc Supplier No. C2342660500X
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trifluoroacetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym Perfluoroacetic Acid, Trifluoroethanoic Acid
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

pH <1
Melting Point -15.4°C
Boiling Point 72.4°C
Color Colorless to Slight Yellow
Quantity 500 mL
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.535
Grade Reagent
Chemical Name or Material Trifluoroacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.