Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, silver salt, 98%

Catalog No. p-41275
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139730050 5 g Glass bottle
AC139730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC139730050 Supplier Thermo Scientific Chemicals Supplier No. 139730050

This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for a quote.

CAS: 2966-50-9 | C2AgF3O2 | 220.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2966-50-9
Molecular Formula C2AgF3O2
Molecular Weight (g/mol) 220.89
MDL Number MFCD00013199
InChI Key KZJPVUDYAMEDRM-UHFFFAOYSA-M
Synonym silver trifluoroacetate, silver 1+ trifluoroacetate, silver i 2,2,2-trifluoroacetate, silver mono trifluoroacetate, trifluoroacetic acid silver salt, silver 1+ ion trifluoroacetate, acetic acid, trifluoro-, silver 1+ salt, silver i trifluoroacetate, trifluoroacetic acid, silver salt, trifluoroacetic acid, silver 1+ salt
PubChem CID 76299
IUPAC Name silver;2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].[Ag+]

Specifications

Melting Point 257.0°C to 260.0°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Glass bottle
Linear Formula CF3CO2Ag
Quantity 5 g
Fieser 01,1018; 07,323; 08,444; 11,471; 16,301; 17,154
Solubility Information Solubility in water: soluble
Formula Weight 220.89
Percent Purity 0.98
Physical Form Crystalline Powder
Chemical Name or Material Trifluoroacetic acid, silver salt, 0.98
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-004-0
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.