Learn More
Medchemexpress LLC Trifluoroacetyl tripeptide-2 | 64577-63-5 | 98.7% | 475.46 | 100 MG
SDP
Catalog No. 5000399185
Encompass_Preferred
Trifluoroacetyl tripeptide-2 is a tripeptide with strong cosmetic activity. It can regulate progerin synthesis, promote extracellular matrix synthesis, and improve skin elasticity. It has anti-wrinkle and firming effects and can be used as an anti-aging ingredient in cosmetic research.
  • Regulates progerin synthesis
  • Promotes extracellular matrix synthesis
  • Improves skin elasticity
  • Provides anti-wrinkle and firming effects
  • Suitable as an anti-aging ingredient in cosmetic research
Catalog No. 50-003-99185 Supplier Medchemexpress LLC Supplier No. HYP5238100MG
Trifluoroacetyl tripeptide-2 is a tripeptide with strong cosmetic activity. It can regulate progerin synthesis, promote extracellular matrix synthesis, and improve skin elasticity. It has anti-wrinkle and firming effects and can be used as an anti-aging ingredient in cosmetic research.
  • Regulates progerin synthesis
  • Promotes extracellular matrix synthesis
  • Improves skin elasticity
  • Provides anti-wrinkle and firming effects
  • Suitable as an anti-aging ingredient in cosmetic research