Learn More
Medchemexpress LLC Trifluoroacetyl tripeptide-2 | 64577-63-5 | 98.71% | 475.46 | 50 MG
SDP
Catalog No. 5000399730
Encompass_Preferred
Trifluoroacetyl tripeptide-2 is a tripeptide with strong cosmetic activity, regulating progerin synthesis and promoting extracellular matrix synthesis. It has anti-wrinkle and firming effects, and can be used as an anti-aging ingredient in cosmetic research.
  • Improves skin elasticity
  • Anti-wrinkle effects
  • Firming effects
  • Suitable as an anti-aging ingredient in cosmetic research
Catalog No. 50-003-99730 Supplier Medchemexpress LLC Supplier No. HYP523850MG
Trifluoroacetyl tripeptide-2 is a tripeptide with strong cosmetic activity, regulating progerin synthesis and promoting extracellular matrix synthesis. It has anti-wrinkle and firming effects, and can be used as an anti-aging ingredient in cosmetic research.
  • Improves skin elasticity
  • Anti-wrinkle effects
  • Firming effects
  • Suitable as an anti-aging ingredient in cosmetic research