Learn More
Cayman Chemical Trifluoroacetyl Tripeptide-2 (acetate), Purity: ≥98%, 250 mg
SDP
Catalog No. 502913635
Encompass_Preferred
A peptide inhibitor of pancreatic and neutrophil elastases (Kis = 0.00056 and 0.5 mM, respectively)
Catalog No. 50-291-3635 Supplier Cayman Chemical Supplier No. 35389250 MG
A peptide inhibitor of pancreatic and neutrophil elastases (Kis = 0.00056 and 0.5 mM, respectively)