Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroethyl Iodide, MP Biomedicals

Catalog No. ICN219313 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219313 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219313 Supplier MP Biomedicals, Inc Supplier No. 0521931305

Chemical Identifiers

CAS 353-83-3
Molecular Formula C2H2F3I
Molecular Weight (g/mol) 209.938
InChI Key RKOUFQLNMRAACI-UHFFFAOYSA-N
Synonym 2-iodo-1,1,1-trifluoroethane, 2,2,2-trifluoroethyl iodide, trifluoroethyl iodide, ethane, 1,1,1-trifluoro-2-iodo, 1-iodo-2,2,2-trifluoroethane, 2,2,2-trifluoroiodoethane, cf3ch2i, unii-ca0j38we9z, 1,1,1-trifluoroiodoethane, 1,1,1-trifluoro-2-iodo-ethane
PubChem CID 67709
IUPAC Name 1,1,1-trifluoro-2-iodoethane
SMILES C(C(F)(F)F)I

Specifications

Quantity 10 g
Formula Weight 209.94
Chemical Name or Material Trifluoroethyl Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.