Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trifluoroethylene

Catalog No. 502908230
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-290-8230 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-290-8230 Supplier Sigma Aldrich Supplier No. 67123125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Trifluoroethylene (trfe) can be used as a monomer unit for the preparation of fluoropolymer (poly(trifluoroethylene)), which finds application in microelectronics, protective/anti-adhesive coatings, chemical industry and li-ion batteries. trfe is also used to synthesize copolymer composites with vinylidene difluoride for various applications.it can also be used to prepare:
  • Pentafluoroethane by reacting with xenon difluoride
  • Lithium (trifluorovinyl)trimethoxy borate by treating with trimethyl borate and n-buli
  • 2,2,3-trifluorooxirane
TRUSTED_SUSTAINABILITY

Specifications

CAS 359-11-5
Molecular Formula C2HF3
Linear Formula FCH=CF2
MDL Number MFCD00039275
Quantity 25 g
Synonym Trifluoroethene
Molecular Weight (g/mol) 82.02
Percent Purity ≥97.5% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.