Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroiodomethane, 99.9%, Thermo Scientific™

Catalog No. AA3966530
Change view
Click to view available options
Quantity:
250 g
Packaging:
Cylinder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA3966530 250 g Cylinder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3966530 Supplier Thermo Scientific Chemicals Supplier No. 3966530

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2314-97-8
Molecular Formula CF3I
Molecular Weight (g/mol) 195.911
MDL Number MFCD00001060
InChI Key VPAYJEUHKVESSD-UHFFFAOYSA-N
Synonym trifluoromethyl iodide, iodotrifluoromethane, methane, trifluoroiodo, perfluoromethyl iodide, trifluoro iodo methane, trifluoromethyliodide, monoiodotrifluoromethane, freon 13t1, unii-42a379kb0u, ccris 7782
PubChem CID 16843
IUPAC Name trifluoro(iodo)methane
SMILES C(F)(F)(F)I

Specifications

Density 2.36g/mL
Boiling Point -22.5°C
Packaging Cylinder
Refractive Index 1.3790
Quantity 250 g
UN Number UN1956
Beilstein 1732740
Sensitivity Light Sensitive
Formula Weight 195.91
Percent Purity 99.9%
Physical Form Gas
Chemical Name or Material Trifluoroiodomethane
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H280-H341
Contains gas under pressure
may explode if heated.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
P281-P201-P308+P313-P405-P410+P403-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Protect from sunlight.
Store in a well-ventilated place.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 2; Proper Shipping Name: COMPRESSED GAS N.O.S.
RTECSNumber PB6975000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.